accessibility menu, dialog, popup

UserName

CH3CH3CH3CH3

CAS RN 3075-84-1

IUPAC Name: 2,2',5,5'-tetramethyl-1,1'-biphenyl
Synonyms: 2,2',5,5'-tetramethyl-1,1'-biphenyl
Molecular Weight (g/mol): 210.32
Molecular Formula: C16H18
InChi Key: ZHTROMYSDSTCCE-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(=C1)C1=CC(C)=CC=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)