missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethylhydrazine, 65 wt% aqueous solution

Catalog No. p-3779747
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC365760050 5 g Glass bottle
AC365760250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC365760050 Supplier Thermo Scientific Chemicals Supplier No. 365760050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5042-30-8 | C2H5F3N2 | 114.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5042-30-8 , 7732-18-5
Molecular Formula C2H5F3N2
Molecular Weight (g/mol) 114.07
MDL Number MFCD00007622
InChI Key OPMFFAOEPFATTG-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyl hydrazine, 2,2,2-trifluoroethylhydrazine, 70 % in aqueous solution, 2,2,2-trifluoroethyl hydrazine in water, sntljqhjptadtauup@, acmc-20aoks, trifluoroethyl hydrazine, cf3ch2nhnh2, 2,2,2-trifluorethylhydrazine
PubChem CID 78740
IUPAC Name 2,2,2-trifluoroethylhydrazine
SMILES C(C(F)(F)F)NN

Specifications

CAS Min % 65.0, 25.0
CAS Max % 75.0, 35.0
Color Colorless to Yellow
Density 1.2940g/mL
Flash Point 42°C
Packaging Glass bottle
Refractive Index 1.3600 to 1.364
Linear Formula CF3CH2NHNH2
Quantity 5 g
Specific Gravity 1.294
Formula Weight 114.07
Percent Purity ≥65%
Physical Form Liquid
Chemical Name or Material 2, 2, 2-Trifluoroethylhydrazine, 70 wt% aqueous solution
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or
EINECSNumber 225-734-

RUO – Research Use Only