accessibility menu, dialog, popup

UserName

CAS RN 5042-30-8

IUPAC Name: (2,2,2-trifluoroethyl)hydrazine
Synonyms: (2,2,2-trifluoroethyl)hydrazine
Molecular Weight (g/mol): 114.07
Molecular Formula: C2H5F3N2
InChi Key: OPMFFAOEPFATTG-UHFFFAOYSA-N
SMILES: NNCC(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results