Learn More
2,2,2-Trifluoroethyl trifluoroacetate, 97+%, Thermo Scientific Chemicals
$80.68 - $80.68
Chemical Identifiers
| CAS | 407-38-5 |
|---|---|
| Molecular Formula | C4H2F6O2 |
| Molecular Weight (g/mol) | 196.05 |
| MDL Number | MFCD00000418 |
| InChI Key | ZKUJOCJJXCPCFS-UHFFFAOYSA-N |
| Synonym | 2,2,2-trifluoroethyl trifluoroacetate, acetic acid, trifluoro-, 2,2,2-trifluoroethyl ester, trifluoroacetic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyltrifluoroacetate, pubchem12623, acmc-1ake7, cf3co2ch2cf3, cf3c o och2cf3, ksc496e0r, trifluoroethyl trifluoroacetate |
| PubChem CID | 67888 |
| IUPAC Name | 2,2,2-trifluoroethyl 2,2,2-trifluoroacetate |
| SMILES | FC(F)(F)COC(=O)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1828314
|
Thermo Scientific Chemicals
A1828314 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 407-38-5 | |
| 196.05 | |
| ZKUJOCJJXCPCFS-UHFFFAOYSA-N | |
| 67888 | |
| FC(F)(F)COC(=O)C(F)(F)F |
| C4H2F6O2 | |
| MFCD00000418 | |
| 2,2,2-trifluoroethyl trifluoroacetate, acetic acid, trifluoro-, 2,2,2-trifluoroethyl ester, trifluoroacetic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyltrifluoroacetate, pubchem12623, acmc-1ake7, cf3co2ch2cf3, cf3c o och2cf3, ksc496e0r, trifluoroethyl trifluoroacetate | |
| 2,2,2-trifluoroethyl 2,2,2-trifluoroacetate |
Specifications
| 407-38-5 | |
| 1.464 | |
| 0°C (32°F) | |
| 1.277 | |
| 25 g | |
| 1783427 | |
| 2,2,2-trifluoroethyl trifluoroacetate, acetic acid, trifluoro-, 2,2,2-trifluoroethyl ester, trifluoroacetic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyltrifluoroacetate, pubchem12623, acmc-1ake7, cf3co2ch2cf3, cf3c o och2cf3, ksc496e0r, trifluoroethyl trifluoroacetate | |
| FC(F)(F)COC(=O)C(F)(F)F | |
| 196.05 | |
| 196.05 | |
| 2,2,2-Trifluoroethyl trifluoroacetate |
| -65°C | |
| 54°C to 55°C | |
| C4H2F6O2 | |
| MFCD00000418 | |
| UN2924 | |
| Moisture sensitive | |
| ZKUJOCJJXCPCFS-UHFFFAOYSA-N | |
| 2,2,2-trifluoroethyl 2,2,2-trifluoroacetate | |
| 67888 | |
| 98% |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber : 206-985-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only