missing translation for 'onlineSavingsMsg'
Learn More

2-(2,2,2-Trifluoroethoxy)aniline 98.0+%, TCI America™

Catalog No. p-7137676
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
T32841G 1 g
T32845G 5 g
2 options

Catalog No. T32841G

Supplier: TCI America T32841G

Only null left

Chemical Identifiers

CAS 57946-60-8
Molecular Formula C8H8F3NO
Molecular Weight (g/mol) 191.153
MDL Number MFCD00077605
InChI Key JIKIVGTUYSNUPQ-UHFFFAOYSA-N
PubChem CID 2777117
IUPAC Name 2-(2,2,2-trifluoroethoxy)aniline
SMILES C1=CC=C(C(=C1)N)OCC(F)(F)F

Specifications

Melting Point 51°C
Quantity 1 g
Formula Weight 191.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2,2,2-Trifluoroethoxy)aniline