accessibility menu, dialog, popup

UserName

CAS RN 57946-60-8

IUPAC Name: 2-(2,2,2-trifluoroethoxy)aniline
Synonyms: 2-(2,2,2-trifluoroethoxy)aniline
Molecular Weight (g/mol): 191.15
Molecular Formula: C8H8F3NO
InChi Key: JIKIVGTUYSNUPQ-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1OCC(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results