missing translation for 'onlineSavingsMsg'
Learn More

2,11-Dithia[3.3]paracyclophane 98.0+%, TCI America™

Catalog No. D2210100MG
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2210100MG 100 mg
D2210500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2210100MG Supplier TCI America Supplier No. D2210100MG

Chemical Identifiers

CAS 28667-63-2
Molecular Formula C16H16S2
Molecular Weight (g/mol) 272.42
MDL Number MFCD00191410
InChI Key CUWSXVJIPZWUGC-UHFFFAOYSA-N
Synonym 3,10-Dithiatricyclo[10.2.2.2(5,8)]octadeca-5,7,12,14,15,17-hexaene
PubChem CID 12238152
IUPAC Name 3,10-dithiatricyclo[10.2.2.2⁵,⁸]octadeca-1(14),5,7,12,15,17-hexaene
SMILES C1SCC2=CC=C(CSCC3=CC=C1C=C3)C=C2

Specifications

Melting Point 222°C
Color White
Quantity 100 mg
Formula Weight 272.42
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,11-Dithia[3.3]paracyclophane