missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Adamantyl)-4-methylphenol, 99%

Catalog No. p-7052809
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5592203 1 g
AAH5592206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5592203 Supplier Thermo Scientific Chemicals Supplier No. H5592203

CAS: 41031-50-9 | C17H22O | 242.36 g/mol

2-(1-Adamantyl)-4-methylphenol, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(1-Adamantyl)-4-methylphenol, is used as a pharmaceutical intermediate.

Solubility
Sparingly Soluble in water (7.8E-3 g/L) (25°C).

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 41031-50-9
Molecular Formula C17H22O
Molecular Weight (g/mol) 242.36
MDL Number MFCD00168147
InChI Key XHLJIHBDAJFXBE-UHFFFAOYSA-N
Synonym 2-1-adamantyl-4-methylphenol, 2-adamantan-1-yl-4-methylphenol, pubchem21690, acmc-1anoj, 2-1-adamantyl-4-methyl-phenol, 2-adamantan-1-yl-4-methyl-phenol, phenol,4-methyl-2-tricyclo 3.3.1.13,7 dec-1-yl, 4-methyl-2-3r,5s,7s-adamantan-1-yl phenol, phenol, 4-methyl-2-tricyclo 3.3.1.1 3,7 dec-1-yl
PubChem CID 617992
IUPAC Name 2-(1-adamantyl)-4-methylphenol
SMILES CC1=CC(=C(O)C=C1)C12CC3CC(CC(C3)C1)C2

Specifications

Melting Point 128°C to 130°C
Color White
Quantity 1 g
Solubility Information Sparingly Soluble in water (7.8E-3g/L) (25°C).
Formula Weight 242.36
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-(1-Adamantyl)-4-methylphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only