missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(-)-Verbenone, ≥99% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

$199.34 - $741.80

Specifications

CAS 1196-01-6
Density 0.975 g/mL (at 20°C)
Flash Point 85°C
Physical Form Neat
Linear Formula C10H14 O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111019009
View Documents
Merck Emd Millipore
948821ML
1 mL
Each for $199.34
Only null left
 
111018118
View Documents
Merck Emd Millipore
948825ML
5 mL
Each for $741.80
Only null left
 
Description

Description

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications

Specifications

1196-01-6
85°C
C10H14 O
227°C to 228°C
Analytical Standard
≥99% (sum of enantiomers, GC)
0.975 g/mL (at 20°C)
Neat
MFCD00065445
(1 S,5 S)-2-Pinen-4-one; (1 S,5 S)-4,6,6-Trimethylbicyclo[3.1.1 ]hept-3-en-2-one
150.22
Analytical Standard
Videos
SDS
Documents

Documents