missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(-)-Verbenone, ≥99% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111019009
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019009 1 mL
111018118 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019009 Supplier Merck Emd Millipore Supplier No. 948821ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 1196-01-6
Density 0.975 g/mL (at 20°C)
Flash Point 85°C
Physical Form Neat
Linear Formula C10H14 O
MDL Number MFCD00065445
Boiling Point 227°C to 228°C
Synonym (1 S,5 S)-2-Pinen-4-one; (1 S,5 S)-4,6,6-Trimethylbicyclo[3.1.1 ]hept-3-en-2-one
Type Analytical Standard
Quantity 1 mL
Formula Weight 150.22
Percent Purity ≥99% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less