Learn More
(1S,2S)-N-(p-Toluenesulfonyl)-1,2-diphenylethanediamine
CAS: 167316-27-0 | C21H23N2O2S | 367.49 g/mol
Supplier: Thermo Scientific Chemicals H2700606
Description
Chiral diamine ligand for cooperative metal-Bronsted acid catalyzed greener reductive amination using hydrogen gas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChiral diamine ligand for cooperative metal-Bronsted acid catalyzed greener reductive amination using hydrogen gas.
Solubility
Solubility in hot Acetonitrile (almost transparency).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
| (1S,2S)-N-(p-Toluenesulfonyl)-1,2-diphenylethanediamine | |
| 127°C to 130°C | |
| MFCD03095684 | |
| 1s,2s-+-n-p-tosyl-1,2-diphenylethylenediamine, s,s-tsdpen, 1s,2s-+-n-4-toluenesulfonyl-1,2-diphenylethylenediamine, 1s,2s-n-4-toluenesulfonyl-1,2-diphenylethylene-1,2-diamine, 1s,2s---n-p-tosyl-1,2-diphenylethylenediamine, n-1s,2s-2-amino-1,2-diphenylethyl-4-methylbenzenesulfonamide, 1s, 2s-+-n-4-toluene sulfonyl 1,2-diphenyl-1,2-ethane diamine, s,s-n-p-toluenesulfonyl-1,2-diphenylethylenediamine, 1s,2s-tsdpen, s,s-n-2-amino-1,2-diphenylethyl-p-toluenesulfonamide | |
| +29° (c=1 in Chloroform) | |
| N-[(1S,2S)-2-amino-1,2-diphenylethyl]-4-methylbenzenesulfonamide | |
| 6612782 |
| 167316-27-0 | |
| C21H23N2O2S | |
| 5 g | |
| UOPFIWYXBIHPIP-SFTDATJTSA-O | |
| CC1=CC=C(C=C1)S(=O)(=O)N[C@H]([C@@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1 | |
| 367.49 | |
| 366.48 |
Chemical Identifiers
| 167316-27-0 | |
| 367.49 | |
| UOPFIWYXBIHPIP-SFTDATJTSA-O | |
| 6612782 | |
| CC1=CC=C(C=C1)S(=O)(=O)N[C@H]([C@@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1 |
| C21H23N2O2S | |
| MFCD03095684 | |
| 1s,2s-+-n-p-tosyl-1,2-diphenylethylenediamine, s,s-tsdpen, 1s,2s-+-n-4-toluenesulfonyl-1,2-diphenylethylenediamine, 1s,2s-n-4-toluenesulfonyl-1,2-diphenylethylene-1,2-diamine, 1s,2s---n-p-tosyl-1,2-diphenylethylenediamine, n-1s,2s-2-amino-1,2-diphenylethyl-4-methylbenzenesulfonamide, 1s, 2s-+-n-4-toluene sulfonyl 1,2-diphenyl-1,2-ethane diamine, s,s-n-p-toluenesulfonyl-1,2-diphenylethylenediamine, 1s,2s-tsdpen, s,s-n-2-amino-1,2-diphenylethyl-p-toluenesulfonamide | |
| N-[(1S,2S)-2-amino-1,2-diphenylethyl]-4-methylbenzenesulfonamide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only