accessibility menu, dialog, popup

UserName

CH3SOONH(S)(S)NH3

CAS RN 167316-27-0

IUPAC Name: N-[(1S,2S)-2-azaniumyl-1,2-diphenylethyl]-4-methylbenzene-1-sulfonamide
Synonyms: N-[(1S,2S)-2-ammonio-1,2-diphenylethyl]-4-methylbenzenesulfonamide
Molecular Weight (g/mol): 367.49
Molecular Formula: C21H23N2O2S
InChi Key: UOPFIWYXBIHPIP-SFTDATJTSA-O
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N[C@H]([C@@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)