missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(+)-2-Benzyloxycyclohexylamine, ChiPros 99+%, ee 99%

Catalog No. AAL1702003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1702003 1 g
AAL1702006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1702003 Supplier Thermo Scientific Chemicals Supplier No. L1702003

CAS: 216394-07-9 | C13H19NO | 205.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 216394-07-9
Molecular Formula C13H19NO
Molecular Weight (g/mol) 205.30
MDL Number MFCD01075753
InChI Key NTHNRYLIXJZHRZ-UHFFFAOYNA-N
PubChem CID 11908838
IUPAC Name (1S,2S)-2-phenylmethoxycyclohexan-1-amine
SMILES NC1CCCCC1OCC1=CC=CC=C1

Specifications

Density 1.02
Boiling Point 79°C to 80°C (0.08 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5275
Quantity 1 g
UN Number UN2735
Beilstein 8981794
Sensitivity Air Sensitive
Synonym 1s,2s-2-benzyloxy cyclohexanamine, 1s,2s-+-2-benzyloxycyclohexylamine, 1s,2s-2-benzyloxycyclohexylamine, 1s,2s-2-benzyloxy cyclohexan-1-amine, 1s,2s-trans-2-benzyloxy-cyclohexylamine, 1s, 2s-2-benzyloxycyclohexylamine, 1s,2s-2-benzyloxy-cyclohexylamine, 1s, 2s-2-benzyloxy-cyclohexylamine, 1s,2s-2-benzyloxy cyclohexylamine, 1s,2s-1-amino-2-benzyloxycyclohexane
Optical Rotation +85° (Neat)
Formula Weight 205.3
Percent Purity ≥99%
Assay ee 99%
Chemical Name or Material (1S,2S)-(+)-2-Benzyloxycyclohexylamine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 606-811-1
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only