accessibility menu, dialog, popup

UserName

NH2O

CAS RN 216394-07-9

IUPAC Name: 2-(benzyloxy)cyclohexan-1-amine
Synonyms: 2-(benzyloxy)cyclohexan-1-amine
Molecular Weight (g/mol): 205.3
Molecular Formula: C13H19NO
InChi Key: NTHNRYLIXJZHRZ-UHFFFAOYNA-N
SMILES: NC1CCCCC1OCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results