missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(+)-2-Benzyloxycyclohexyl isothiocyanate, 97%

Catalog No. AAL2006003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2006003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL2006003 Supplier Thermo Scientific Chemicals Supplier No. L2006003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 737000-89-4 | C14H17NOS | 247.356 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 737000-89-4
Molecular Formula C14H17NOS
Molecular Weight (g/mol) 247.356
MDL Number MFCD05664047
InChI Key DNRPSBSMXLYTQF-KBPBESRZSA-N
Synonym 1s,2s-+-2-benzyloxycyclohexyl isothiocyanate, 1s,2s-2-isothiocyanatocyclohexyl oxy methyl benzene, 1s,2s-2-phenylmethoxy cyclohexanisothiocyanate
PubChem CID 11863573
IUPAC Name [(1S,2S)-2-isothiocyanatocyclohexyl]oxymethylbenzene
SMILES C1CCC(C(C1)N=C=S)OCC2=CC=CC=C2

Specifications

Melting Point 33°C to 36°C
Density 1.106
Refractive Index 1.5674
Quantity 1 g
UN Number UN2923
Sensitivity Moisture sensitive
Formula Weight 247.36
Percent Purity 97%
Chemical Name or Material (1S,2S)-(+)-2-Benzyloxycyclohexyl isothiocyanate
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only