CAS RN 737000-89-4
IUPAC Name:
{[(2-isothiocyanatocyclohexyl)oxy]methyl}benzene
Synonyms:
{[(2-isothiocyanatocyclohexyl)oxy]methyl}benzene
Molecular Weight (g/mol):
247.36
Molecular Formula:
C14H17NOS
InChi Key:
DNRPSBSMXLYTQF-UHFFFAOYNA-N
SMILES:
S=C=NC1CCCCC1OCC1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
(1S,2S)-(+)-2-Benzyloxycyclohexyl isothiocyanate, 97%
(1S,2S)-(+)-2-Benzyloxycyclohexyl isothiocyanate, 97%
CAS: 737000-89-4 Molecular Formula: C14H17NOS Molecular Weight (g/mol): 247.356 MDL Number: MFCD05664047 InChI Key: DNRPSBSMXLYTQF-KBPBESRZSA-N Synonym: 1s,2s-+-2-benzyloxycyclohexyl isothiocyanate,1s,2s-2-isothiocyanatocyclohexyl oxy methyl benzene,1s,2s-2-phenylmethoxy cyclohexanisothiocyanate PubChem CID: 11863573 IUPAC Name: [(1S,2S)-2-isothiocyanatocyclohexyl]oxymethylbenzene SMILES: C1CCC(C(C1)N=C=S)OCC2=CC=CC=C2