missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-2-Amino-1-phenylpropyldiphenylphosphine, 97%, Thermo Scientific™

Catalog No. p-4229067
Change view
Click to view available options
Quantity:
100 mg
250 mg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434141000 100 mg Glass bottle
AC434142500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434141000 Supplier Thermo Scientific Chemicals Supplier No. 434141000
Only null left

Chemical Identifiers

CAS 341968-71-6
Molecular Formula C21H22NP
Molecular Weight (g/mol) 319.39
MDL Number MFCD11044864
InChI Key JWZAIGGNEGTDMG-AURADERDNA-N
Synonym 1s,2s-1-diphenylphosphino-1-phenylpropan-2-amine, 1s,2s-2-amino-1-phenylpropyl diphenylphosphine, 1s,2s-2-amino-1-phenylpropyldiphenylphosphine, 1s,2s-1-diphenylphosphanyl-1-phenylpropan-2-amine, 1s,2s-2-diphenylphosphino-1-methyl-2-phenylethylamine, 1s,2s-2-amino-1-phenylpropyl diphenylphosphane, 1s,2s-1-diphenylphosphino-1-phenyl-2-propanamine, 1s,2s-2-amino-1-phenylpropyldiphenylphosphine,min, s,s-+-2-amino-1-phenylpropyldiphenylphosphine, 1s,2s-2-amino-1-phenylpropyl diphenylphosphine, kanata purity
PubChem CID 46176707
IUPAC Name (1S,2S)-1-diphenylphosphanyl-1-phenylpropan-2-amine
SMILES C[C@H](N)[C@@H](P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 100.0°C to 105.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 mg
Formula Weight 319.38
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (1S, 2S)-2-Amino-1-phenylpropyldiphenylphosphine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only