accessibility menu, dialog, popup

UserName

CAS RN 341968-71-6

IUPAC Name: [(1S,2S)-2-amino-1-phenylpropyl]diphenylphosphane
Synonyms: [(1S,2S)-2-amino-1-phenylpropyl]diphenylphosphane
Molecular Weight (g/mol): 319.39
Molecular Formula: C21H22NP
InChi Key: JWZAIGGNEGTDMG-AURADERDNA-N
SMILES: C[C@H](N)[C@@H](P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results