missing translation for 'onlineSavingsMsg'
Learn More

(1S,2R)-2-[N-Benzyl-N-(mesitylenesulfonyl)amino]-1-phenyl-1-propanol 98.0+%, TCI America™

Catalog No. B21041G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21041G 1 g
B21045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21041G Supplier TCI America Supplier No. B21041G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 187324-64-7
Molecular Formula C25H29NO3S
Molecular Weight (g/mol) 423.571
MDL Number MFCD02093447
InChI Key ZPERKXJNXPLVAC-ZJSXRUAMSA-N
Synonym (1S,2R)-N-Benzyl-N-(mesitylenesulfonyl)norephedrine, (1S,2R)-2-[N-Benzyl-N-(2,4,6-trimethylbenzenesulfonyl)amino]-1-phenyl-1-propanol
PubChem CID 44630156
IUPAC Name N-benzyl-N-[(1S,2R)-1-hydroxy-1-phenylpropan-2-yl]-2,4,6-trimethylbenzenesulfonamide
SMILES CC1=CC(=C(C(=C1)C)S(=O)(=O)N(CC2=CC=CC=C2)C(C)C(C3=CC=CC=C3)O)C

Specifications

Color White
Quantity 1 g
Formula Weight 423.57
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (1S,2R)-2-[N-Benzyl-N-(mesitylenesulfonyl)amino]-1-phenyl-1-propanol