accessibility menu, dialog, popup

UserName

CH3H3CSOONH3COHCH3

CAS RN 187324-64-7

IUPAC Name: N-benzyl-N-(1-hydroxy-1-phenylpropan-2-yl)-2,4,6-trimethylbenzene-1-sulfonamide
Synonyms: N-benzyl-N-(1-hydroxy-1-phenylpropan-2-yl)-2,4,6-trimethylbenzenesulfonamide
Molecular Weight (g/mol): 423.57
Molecular Formula: C25H29NO3S
InChi Key: ZPERKXJNXPLVAC-UHFFFAOYNA-N
SMILES: CC(C(O)C1=CC=CC=C1)N(CC1=CC=CC=C1)S(=O)(=O)C1=C(C)C=C(C)C=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results