missing translation for 'onlineSavingsMsg'
Learn More

(1S,2R)-2-(Dibutylamino)-1-phenyl-1-propanol 98.0+%, TCI America™

Catalog No. p-7127122
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21291G 1 g
D21295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21291G Supplier TCI America Supplier No. D21291G

Chemical Identifiers

CAS 114389-70-7
Molecular Formula C17H29NO
Molecular Weight (g/mol) 263.425
MDL Number MFCD00142695
InChI Key BRRGNOFUBFINSX-NVXWUHKLSA-N
Synonym (-)-alpha-[1-(Dibutylamino)ethyl]benzyl Alcohol, (-)-N,N-Dibutylnorephedrin
PubChem CID 11003877
IUPAC Name (1S,2R)-2-(dibutylamino)-1-phenylpropan-1-ol
SMILES CCCCN(CCCC)C(C)C(C1=CC=CC=C1)O

Specifications

Color Yellow
Boiling Point 170°C
Quantity 1 g
Formula Weight 263.43
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (1S,2R)-2-(Dibutylamino)-1-phenyl-1-propanol