missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1R)-(+)-alpha-Pinene 97.0+%, TCI America™
$55.07 - $360.74
Chemical Identifiers
| CAS | 7785-70-8 |
|---|---|
| Molecular Formula | C10H16 |
| Molecular Weight (g/mol) | 136.24 |
| MDL Number | MFCD00001346 |
| InChI Key | GRWFGVWFFZKLTI-RKDXNWHRSA-N |
| Synonym | +-alpha-pinene, 1r,5r-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, 1r-+-alpha-pinene, 1r,5r-alpha-pinene, unii-h6cm4twh1w, +-pinene, h6cm4twh1w, 1r,5r-2-pinene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1r,5r, +-pin-2 3-ene |
| PubChem CID | 82227 |
| ChEBI | CHEBI:28261 |
| IUPAC Name | (1R,5R)-2,6,6-trimethylbicyclo[3.1.1]hept-2-ene |
| SMILES | CC1=CC[C@@H]2C[C@H]1C2(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
P109925ML
|
N/A | 25 mL |
Each for $55.07
|
|
|||||
|
P1099500ML
|
N/A | 500 mL |
Each for $360.74
|
|
|||||
Chemical Identifiers
| C10H16 | |
| MFCD00001346 | |
| +-alpha-pinene, 1r,5r-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, 1r-+-alpha-pinene, 1r,5r-alpha-pinene, unii-h6cm4twh1w, +-pinene, h6cm4twh1w, 1r,5r-2-pinene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1r,5r, +-pin-2 3-ene | |
| CHEBI:28261 | |
| CC1=CC[C@@H]2C[C@H]1C2(C)C |
Specifications
| 7785-70-8 | |
| 156°C | |
| MFCD00001346 | |
| +-alpha-pinene, 1r,5r-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, 1r-+-alpha-pinene, 1r,5r-alpha-pinene, unii-h6cm4twh1w, +-pinene, h6cm4twh1w, 1r,5r-2-pinene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1r,5r, +-pin-2 3-ene | |
| CC1=CC[C@@H]2C[C@H]1C2(C)C | |
| 136.24 | |
| CHEBI:28261 | |
| ≥97.0% (GC) | |
| (1R)-(+)-alpha-Pinene |
| Colorless | |
| C10H16 | |
| 2368 | |
| GRWFGVWFFZKLTI-RKDXNWHRSA-N | |
| (1R,5R)-2,6,6-trimethylbicyclo[3.1.1]hept-2-ene | |
| 82227 | |
| 136.24 | |
| Liquid |
Safety and Handling
EINECSNumber : (4)-0593&(8)-0497
TSCA : Yes