missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1R,2R)-(+)-1,2-Cyclohexanediamine L-Tartrate 98.0+%, TCI America™
Supplier: TCI America C14495G
Specifications
| (1R,2R)-(+)-1,2-Cyclohexanediamine L-Tartrate | |
| 273°C | |
| C10H20N2O6 | |
| 5 g | |
| GDOTUTAQOJUZOF-ZXZVGZDWSA-N | |
| (1R,2R)-cyclohexane-1,2-diamine;(2R,3R)-2,3-dihydroxybutanedioic acid | |
| 11448443 | |
| ≥98.0% (T) |
| 39961-95-0 | |
| White | |
| MFCD00191979 | |
| 1r,2r-+-1,2-diaminocyclohexane l-tartrate, 1r,2r-cyclohexane-1,2-diamine 2r,3r-2,3-dihydroxysuccinate, 1r,2r-+-1,2-cyclohexanediamine l-tartrate, 1r,2r---cyclohexane-1,2-diamine l-tartrate salt, 1r-trans-1,2-diaminocyclohexane l-tartrate, 1r,2r-+-cyclohexane-1,2-diamine l-tartrate, 1r,2r-1,2-diaminocyclohexane l-tartrate, 1r,2r-cyclohexane-1,2-diamine; l +-tartaric acid, pubchem17364, ksc221k4n | |
| C1CCC(C(C1)N)N.C(C(C(=O)O)O)(C(=O)O)O | |
| 264.278 | |
| 264.28 | |
| Crystalline Powder |
Chemical Identifiers
| 39961-95-0 | |
| 264.278 | |
| GDOTUTAQOJUZOF-ZXZVGZDWSA-N | |
| 11448443 | |
| C1CCC(C(C1)N)N.C(C(C(=O)O)O)(C(=O)O)O |
| C10H20N2O6 | |
| MFCD00191979 | |
| 1r,2r-+-1,2-diaminocyclohexane l-tartrate, 1r,2r-cyclohexane-1,2-diamine 2r,3r-2,3-dihydroxysuccinate, 1r,2r-+-1,2-cyclohexanediamine l-tartrate, 1r,2r---cyclohexane-1,2-diamine l-tartrate salt, 1r-trans-1,2-diaminocyclohexane l-tartrate, 1r,2r-+-cyclohexane-1,2-diamine l-tartrate, 1r,2r-1,2-diaminocyclohexane l-tartrate, 1r,2r-cyclohexane-1,2-diamine; l +-tartaric acid, pubchem17364, ksc221k4n | |
| (1R,2R)-cyclohexane-1,2-diamine;(2R,3R)-2,3-dihydroxybutanedioic acid |
Safety and Handling
TSCA : No