missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-(+)-1,2-Cyclohexanediamine L-Tartrate 98.0+%, TCI America™

Catalog No. p-7124989
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14495G 5 g
C144925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14495G Supplier TCI America Supplier No. C14495G
Only null left

Chemical Identifiers

CAS 39961-95-0
Molecular Formula C10H20N2O6
Molecular Weight (g/mol) 264.278
MDL Number MFCD00191979
InChI Key GDOTUTAQOJUZOF-ZXZVGZDWSA-N
Synonym 1r,2r-+-1,2-diaminocyclohexane l-tartrate, 1r,2r-cyclohexane-1,2-diamine 2r,3r-2,3-dihydroxysuccinate, 1r,2r-+-1,2-cyclohexanediamine l-tartrate, 1r,2r---cyclohexane-1,2-diamine l-tartrate salt, 1r-trans-1,2-diaminocyclohexane l-tartrate, 1r,2r-+-cyclohexane-1,2-diamine l-tartrate, 1r,2r-1,2-diaminocyclohexane l-tartrate, 1r,2r-cyclohexane-1,2-diamine; l +-tartaric acid, pubchem17364, ksc221k4n
PubChem CID 11448443
IUPAC Name (1R,2R)-cyclohexane-1,2-diamine;(2R,3R)-2,3-dihydroxybutanedioic acid
SMILES C1CCC(C(C1)N)N.C(C(C(=O)O)O)(C(=O)O)O

Specifications

Melting Point 273°C
Color White
Quantity 5 g
Formula Weight 264.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R)-(+)-1,2-Cyclohexanediamine L-Tartrate