accessibility menu, dialog, popup

UserName

CAS RN 39961-95-0

IUPAC Name: 2,3-dihydroxybutanedioic acid; cyclohexane-1,2-diamine
Synonyms: (.+-.)-tartaric acid; 1,2-cyclohexanediamine
Molecular Weight (g/mol): 264.28
Molecular Formula: C10H20N2O6
InChi Key: GDOTUTAQOJUZOF-UHFFFAOYNA-N
SMILES: NC1CCCCC1N.OC(C(O)C(O)=O)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)