missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H,1H,2H,2H-Tridecafluoro-n-octyl Iodide 97.0+%, TCI America™
Supplier: TCI America T207425G
Specifications
| 1H,1H,2H,2H-Tridecafluoro-n-octyl Iodide | |
| Yellow | |
| C8H4F13I | |
| 25 g | |
| NVVZEKTVIXIUKW-UHFFFAOYSA-N | |
| 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-8-iodooctane | |
| 74888 | |
| ≥97.0% (GC) |
| 2043-57-4 | |
| 180°C | |
| MFCD00039410 | |
| 1h,1h,2h,2h-perfluorooctyl iodide, octane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-8-iodo, 1-iodo-1h,1h,2h,2h-perfluorooctane, 1,1,2,2-tetrahydroperfluorooctyliodide, 1,1,2,2-tetrahydroperfluorooctyl iodide, 2-perfluorohexyl ethyl iodide, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-iodooctane, 1h,1h,2h,2h-tridecafluoro-n-octyl iodide, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl iodide, acmc-1ccsn | |
| C(CI)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F | |
| 474.004 | |
| 474.00 | |
| Liquid |
Chemical Identifiers
| 2043-57-4 | |
| 474.004 | |
| NVVZEKTVIXIUKW-UHFFFAOYSA-N | |
| 74888 | |
| C(CI)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F |
| C8H4F13I | |
| MFCD00039410 | |
| 1h,1h,2h,2h-perfluorooctyl iodide, octane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-8-iodo, 1-iodo-1h,1h,2h,2h-perfluorooctane, 1,1,2,2-tetrahydroperfluorooctyliodide, 1,1,2,2-tetrahydroperfluorooctyl iodide, 2-perfluorohexyl ethyl iodide, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-iodooctane, 1h,1h,2h,2h-tridecafluoro-n-octyl iodide, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl iodide, acmc-1ccsn | |
| 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-8-iodooctane |
Safety and Handling
EINECSNumber : (2)-4010
TSCA : Yes