accessibility menu, dialog, popup

UserName

CAS RN 2043-57-4

IUPAC Name: 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-8-iodooctane
Synonyms: 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-8-iodooctane
Molecular Weight (g/mol): 474
Molecular Formula: C8H4F13I
InChi Key: NVVZEKTVIXIUKW-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCI
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)