missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Tridecafluoro-n-octyl Iodide 97.0+%, TCI America™

Catalog No. p-7137089
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20745G 5 g
T207425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20745G Supplier TCI America Supplier No. T20745G
Only null left

Chemical Identifiers

CAS 2043-57-4
Molecular Formula C8H4F13I
Molecular Weight (g/mol) 474.004
MDL Number MFCD00039410
InChI Key NVVZEKTVIXIUKW-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorooctyl iodide, octane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-8-iodo, 1-iodo-1h,1h,2h,2h-perfluorooctane, 1,1,2,2-tetrahydroperfluorooctyliodide, 1,1,2,2-tetrahydroperfluorooctyl iodide, 2-perfluorohexyl ethyl iodide, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-iodooctane, 1h,1h,2h,2h-tridecafluoro-n-octyl iodide, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl iodide, acmc-1ccsn
PubChem CID 74888
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-8-iodooctane
SMILES C(CI)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Yellow
Boiling Point 180°C
Quantity 5 g
Formula Weight 474.00
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1H,1H,2H,2H-Tridecafluoro-n-octyl Iodide
EINECSNumber (2)-4010
TSCA Yes