missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H-1,2,3-Triazole 98.0+%, TCI America™
Supplier: TCI America T188025G
Specifications
| 1H-1,2,3-Triazole | |
| White-Yellow | |
| MFCD00014490 | |
| 1h-1,2,3-triazole, 1,2,3-triazole, 2h-1,2,3-triazole, 1,2,3-1h-triazole, triazole, triazol, osotriazole, v-triazole, 1,2,3-triazol, 1,2,3 triazole | |
| C1=NNN=C1 | |
| 69.067 | |
| CHEBI:35565 | |
| ≥98.0% (T) |
| 288-36-8 | |
| C2H3N3 | |
| 25 g | |
| QWENRTYMTSOGBR-UHFFFAOYSA-N | |
| 2H-triazole | |
| 67516 | |
| 69.07 | |
| Crystalline Lumps |
Chemical Identifiers
| 288-36-8 | |
| 69.067 | |
| QWENRTYMTSOGBR-UHFFFAOYSA-N | |
| 67516 | |
| 2H-triazole |
| C2H3N3 | |
| MFCD00014490 | |
| 1h-1,2,3-triazole, 1,2,3-triazole, 2h-1,2,3-triazole, 1,2,3-1h-triazole, triazole, triazol, osotriazole, v-triazole, 1,2,3-triazol, 1,2,3 triazole | |
| CHEBI:35565 | |
| C1=NNN=C1 |
Safety and Handling
EINECSNumber : (5)-6196
TSCA : No