accessibility menu, dialog, popup

UserName

HNNN

CAS RN 288-36-8

IUPAC Name: 2H-1,2,3-triazole
Synonyms: 2H-1,2,3-triazole osotriazole
Molecular Weight (g/mol): 69.07
Molecular Formula: C2H3N3
InChi Key: QWENRTYMTSOGBR-UHFFFAOYSA-N
SMILES: N1N=CC=N1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)