missing translation for 'onlineSavingsMsg'
Learn More

1H-1,2,3-Triazole 98.0+%, TCI America™

Catalog No. p-7137000
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18805G 5 g
T188025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18805G Supplier TCI America Supplier No. T18805G
Only null left

Chemical Identifiers

CAS 288-36-8
Molecular Formula C2H3N3
Molecular Weight (g/mol) 69.067
MDL Number MFCD00014490
InChI Key QWENRTYMTSOGBR-UHFFFAOYSA-N
Synonym 1h-1,2,3-triazole, 1,2,3-triazole, 2h-1,2,3-triazole, 1,2,3-1h-triazole, triazole, triazol, osotriazole, v-triazole, 1,2,3-triazol, 1,2,3 triazole
PubChem CID 67516
ChEBI CHEBI:35565
IUPAC Name 2H-triazole
SMILES C1=NNN=C1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 69.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material 1H-1,2,3-Triazole
EINECSNumber (5)-6196
TSCA No