missing translation for 'onlineSavingsMsg'
Learn More
Learn More
16-Hydroxyhexadecanoic Acid 98.0+%, TCI America™
$146.20 - $719.57
Chemical Identifiers
| CAS | 506-13-8 |
|---|---|
| Molecular Formula | C16H32O3 |
| Molecular Weight (g/mol) | 272.429 |
| MDL Number | MFCD00002750 |
| InChI Key | UGAGPNKCDRTDHP-UHFFFAOYSA-N |
| Synonym | juniperic acid, 16-hydroxypalmitic acid, hexadecanoic acid, 16-hydroxy, palmitic acid, 16-hydroxy, omega-hydroxypalmitic acid, 16-hydroxy-hexadecanoic acid, unii-7ipp3u0f3i, .omega.-hydroxypalmitic acid, 7ipp3u0f3i, 16-hydroxy hexadecanoic acid |
| PubChem CID | 10466 |
| ChEBI | CHEBI:55328 |
| IUPAC Name | 16-hydroxyhexadecanoic acid |
| SMILES | C(CCCCCCCC(=O)O)CCCCCCCO |
Chemical Identifiers
| C16H32O3 | |
| MFCD00002750 | |
| juniperic acid, 16-hydroxypalmitic acid, hexadecanoic acid, 16-hydroxy, palmitic acid, 16-hydroxy, omega-hydroxypalmitic acid, 16-hydroxy-hexadecanoic acid, unii-7ipp3u0f3i, .omega.-hydroxypalmitic acid, 7ipp3u0f3i, 16-hydroxy hexadecanoic acid | |
| CHEBI:55328 | |
| C(CCCCCCCC(=O)O)CCCCCCCO |
Specifications
| 506-13-8 | |
| C16H32O3 | |
| juniperic acid, 16-hydroxypalmitic acid, hexadecanoic acid, 16-hydroxy, palmitic acid, 16-hydroxy, omega-hydroxypalmitic acid, 16-hydroxy-hexadecanoic acid, unii-7ipp3u0f3i, .omega.-hydroxypalmitic acid, 7ipp3u0f3i, 16-hydroxy hexadecanoic acid | |
| C(CCCCCCCC(=O)O)CCCCCCCO | |
| 272.429 | |
| CHEBI:55328 | |
| ≥98.0% (T) | |
| 16-Hydroxyhexadecanoic Acid |
| 99°C | |
| MFCD00002750 | |
| UGAGPNKCDRTDHP-UHFFFAOYSA-N | |
| 16-hydroxyhexadecanoic acid | |
| 10466 | |
| 272.43 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1340
TSCA : Yes