missing translation for 'onlineSavingsMsg'
Learn More

16-Hydroxyhexadecanoic Acid 98.0+%, TCI America™

Catalog No. H06751G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06751G 1 g
H06755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H06751G Supplier TCI America Supplier No. H06751G

Chemical Identifiers

CAS 506-13-8
Molecular Formula C16H32O3
Molecular Weight (g/mol) 272.429
MDL Number MFCD00002750
InChI Key UGAGPNKCDRTDHP-UHFFFAOYSA-N
Synonym juniperic acid, 16-hydroxypalmitic acid, hexadecanoic acid, 16-hydroxy, palmitic acid, 16-hydroxy, omega-hydroxypalmitic acid, 16-hydroxy-hexadecanoic acid, unii-7ipp3u0f3i, .omega.-hydroxypalmitic acid, 7ipp3u0f3i, 16-hydroxy hexadecanoic acid
PubChem CID 10466
ChEBI CHEBI:55328
IUPAC Name 16-hydroxyhexadecanoic acid
SMILES C(CCCCCCCC(=O)O)CCCCCCCO

Specifications

Melting Point 99°C
Quantity 1 g
Formula Weight 272.43
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 16-Hydroxyhexadecanoic Acid
EINECSNumber (2)-1340
TSCA Yes