accessibility menu, dialog, popup

UserName

HOOHO

CAS RN 506-13-8

IUPAC Name: 16-hydroxyhexadecanoic acid
Synonyms: 16-hydroxyhexadecanoic acid juniperic acid ω-hydroxypalmitic acid
Molecular Weight (g/mol): 272.43
Molecular Formula: C16H32O3
InChi Key: UGAGPNKCDRTDHP-UHFFFAOYSA-N
SMILES: OCCCCCCCCCCCCCCCC(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)