missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 1500 Series Class II Type B2 Bio Safety Cabinets
Our Class II, Type B2 Biological Safety Cabinet is designed for applications that require greater dilution of volatile toxic chemicals or the aseptic processing of hazardous drugs.
$77,551.85 - $89,263.49
Specifications
| Certifications/Compliance | NSF Class II Type B2 BSC, NSF/ANSI 49, UL, CE |
|---|---|
| Chamber Material | Stainless Steel (304) |
| Product Type | Biosafety Cabinet |
| Model | 1500 Series B2 |
| Sash Opening (Metric) | 20.3 cm |
| Catalog Number | Mfr. No. | Plug Type | Width (English) Cabinet | Dimensions (L x W x H) Exterior | Lighting | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Plug Type | Width (English) Cabinet | Dimensions (L x W x H) Exterior | Lighting | Price | Quantity & Availability | ||
|
13201871
|
Thermo Scientific™
1512B2 |
North America (115 V, 15 A) | 6 ft. | 31.6 x 78 x 72.6 in. | Without UV light |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
13201866
|
Thermo Scientific™
1502B2 |
North America (115 V, 15 A) | 4 ft. | 31.6 x 54.3 x 72.6 in. | With UV light |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
13201862
|
Thermo Scientific™
1519B2 |
North America (115 V, 15 A) | 6 ft. | 31.6 x 78 x 72.6 in. | With UV light |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Thermo Scientific™ 1500 Series Class II, Type B2 BSC is available in nominal widths of 4 ft (1.2 m) and 6 ft (1.8 m) and is fully compliant with NSF/ANSI 49 standards. It features an ergonomic design, advanced DC motor technology, and a convenient touchscreen user interface allows for easy monitoring of cabinet conditions.
- NSF/ANSI 49 listed Class II, Type B2 BSC
- Auto compensation for downflow, allowing for safe use of air pre-filters for additional extension of HEPA filter life
- Advanced DC motor
- Stainless steel interior
- 5 in. touchscreen display for visualizing and monitoring cabinet conditions, including, airflow status, alerts and alarms, UV light timer, HEPA filter life indicator, Data logging, and Connectivity
Specifications
| NSF Class II Type B2 BSC, NSF/ANSI 49, UL, CE | |
| Biosafety Cabinet | |
| 20.3 cm |
| Stainless Steel (304) | |
| 1500 Series B2 | |
| 8 in. |