Learn More
Thermo Scientific™ 1500 Series Class II Type B2 Bio Safety Cabinets
Our Class II, Type B2 Biological Safety Cabinet is designed for applications that require greater dilution of volatile toxic chemicals or the aseptic processing of hazardous drugs.
Supplier: Thermo Scientific™ 1502B2
Description
Thermo Scientific™ 1500 Series Class II, Type B2 BSC is available in nominal widths of 4 ft (1.2 m) and 6 ft (1.8 m) and is fully compliant with NSF/ANSI 49 standards. It features an ergonomic design, advanced DC motor technology, and a convenient touchscreen user interface allows for easy monitoring of cabinet conditions.
- NSF/ANSI 49 listed Class II, Type B2 BSC
- Auto compensation for downflow, allowing for safe use of air pre-filters for additional extension of HEPA filter life
- Advanced DC motor
- Stainless steel interior
- 5 in. touchscreen display for visualizing and monitoring cabinet conditions, including, airflow status, alerts and alarms, UV light timer, HEPA filter life indicator, Data logging, and Connectivity
Specifications
| NSF Class II Type B2 BSC, NSF/ANSI 49, UL, CE | |
| Biosafety Cabinet | |
| 65 dBA | |
| 2 Duplex, GFI | |
| 54.3 in. | |
| With UV light | |
| 31.6 x 54.3 x 72.6 in. | |
| 4 ft. | |
| 115 V |
| Stainless Steel (304) | |
| 1500 Series B2 | |
| North America (115 V, 15 A) | |
| 20.3 cm | |
| 137.9 cm | |
| 8 in. | |
| 48.5 in. | |
| 115 V, 50/60 Hz | |
| 0.28 kW |