missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(11S,12S)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine 98.0+%, TCI America™
$211.77 - $1099.28
Chemical Identifiers
| CAS | 138517-66-5 |
|---|---|
| Molecular Formula | C16H16N2 |
| Molecular Weight (g/mol) | 236.318 |
| MDL Number | MFCD06654768 |
| InChI Key | NWDYSRZJOLDMRE-CKUJCDMFSA-N |
| PubChem CID | 11499644 |
| SMILES | C1=CC=C2C3C(C(C(C2=C1)C4=CC=CC=C34)N)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D3445100MG
|
TCI America
D3445100MG |
100 mg |
Each for $211.77
|
|
|||||
|
D34451G
|
TCI America
D34451G |
1 g |
Each for $1,099.28
|
|
|||||
Chemical Identifiers
| 138517-66-5 | |
| 236.318 | |
| NWDYSRZJOLDMRE-CKUJCDMFSA-N | |
| C1=CC=C2C3C(C(C(C2=C1)C4=CC=CC=C34)N)N |
| C16H16N2 | |
| MFCD06654768 | |
| 11499644 |
Specifications
| 138517-66-5 | |
| White-Yellow | |
| MFCD06654768 | |
| NWDYSRZJOLDMRE-CKUJCDMFSA-N | |
| 236.318 | |
| 236.32 | |
| Crystalline Powder |
| 158°C | |
| C16H16N2 | |
| 100 mg | |
| C1=CC=C2C3C(C(C(C2=C1)C4=CC=CC=C34)N)N | |
| 11499644 | |
| ≥98.0% (HPLC,T) | |
| (11S,12S)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine |
Safety and Handling
TSCA : No