missing translation for 'onlineSavingsMsg'
Learn More

(11S,12S)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine 98.0+%, TCI America™

Catalog No. d3445100mg
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
100 mg
Catalog No. Quantity
D3445100MG 100 mg
D34451G 1 g
2 options

Catalog No. D3445100MG

Supplier: TCI America D3445100MG

Only null left

Chemical Identifiers

CAS 138517-66-5
Molecular Formula C16H16N2
Molecular Weight (g/mol) 236.318
MDL Number MFCD06654768
InChI Key NWDYSRZJOLDMRE-CKUJCDMFSA-N
PubChem CID 11499644
SMILES C1=CC=C2C3C(C(C(C2=C1)C4=CC=CC=C34)N)N

Specifications

Melting Point 158°C
Color White-Yellow
Quantity 100 mg
Formula Weight 236.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (11S,12S)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine