missing translation for 'onlineSavingsMsg'
Learn More

11-Bromo-1-undecene, 90+%

Catalog No. p-7026063
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2184906 5 g
AAB2184914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2184906 Supplier Thermo Scientific Chemicals Supplier No. B2184906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7766-50-9 | C11H21Br | 233.193 g/mol

11-Bromo-1-undecene is used in advanced polymer dielectrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
11-Bromo-1-undecene is used in advanced polymer dielectrics.

Solubility
Soluble in water, 0.2422 mg/L @ 25°C (est).

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 7766-50-9
Molecular Formula C11H21Br
Molecular Weight (g/mol) 233.193
MDL Number MFCD00040825
InChI Key YPLVPFUSXYSHJD-UHFFFAOYSA-N
Synonym 11-bromo-1-undecene, 10-undecenyl bromide, 1-bromo-10-undecene, undec-10-enyl bromide, 11-bromo-undec-1-ene, 1-undecene, 11-bromo, 11-bromoundecene, undecylenyl bromide, n-undecylenic bromide, w-undecylenyl bromide
PubChem CID 284148
IUPAC Name 11-bromoundec-1-ene
SMILES C=CCCCCCCCCCBr

Specifications

Density 1.063
Boiling Point 149°C to 150°C (35 mmHg)
Flash Point >110°C (230°F)
Assay Percent Range 0.96
Refractive Index 1.469
Quantity 5 g
Beilstein 1753231
Solubility Information Soluble in water,0.2422mg/L @ 25°C (est).
Formula Weight 233.2
Percent Purity 96%
Chemical Name or Material 11-Bromo-1-undecene
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only