missing translation for 'onlineSavingsMsg'
Learn More

11-Azidoundecyltriethoxysilane, 95%

CAS: 663171-33-3 | C17H37N3O3Si | 359.586 g/mol

$133.11 - $358.36

Chemical Identifiers

CAS 663171-33-3
Molecular Formula C17H37N3O3Si
Molecular Weight (g/mol) 359.586
MDL Number MFCD11982879
InChI Key WCDFLYKXACDAES-UHFFFAOYSA-N
Synonym azidoundecyltriethoxysilane, 11-azidoundecyltriethoxysilane, 11-azidoundecyl triethoxysilane
PubChem CID 46779111
IUPAC Name 11-azidoundecyl(triethoxy)silane
SMILES CCO[Si](CCCCCCCCCCCN=[N+]=[N-])(OCC)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH33524MD
View Documents
Thermo Scientific Chemicals
H33524MD
250 mg
Each for $133.11
Only null left
 
AAH3352403
View Documents
Thermo Scientific Chemicals
H3352403
1 g
Each for $358.36
Only null left
 
Description

Description

This coupling agent specifically reacts with alkyne functions to form the corresponding 1,2,3-triazole via a copper-free 1,3-dipolar cycloaddition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This coupling agent specifically reacts with alkyne functions to form the corresponding 1,2,3-triazole via a copper-free 1,3-dipolar cycloaddition.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

663171-33-3
359.586
WCDFLYKXACDAES-UHFFFAOYSA-N
46779111
CCO[Si](CCCCCCCCCCCN=[N+]=[N-])(OCC)OCC
C17H37N3O3Si
MFCD11982879
azidoundecyltriethoxysilane, 11-azidoundecyltriethoxysilane, 11-azidoundecyl triethoxysilane
11-azidoundecyl(triethoxy)silane

Specifications

110°C to 115°C (0.04mmHg)
250 mg
N3(CH2)11Si(OCH2CH3)3
azidoundecyltriethoxysilane, 11-azidoundecyltriethoxysilane, 11-azidoundecyl triethoxysilane
CCO[Si](CCCCCCCCCCCN=[N+]=[N-])(OCC)OCC
359.586
359.58
1.4390-1.4430
11-Azidoundecyltriethoxysilane
663171-33-3
C17H37N3O3Si
MFCD11982879
WCDFLYKXACDAES-UHFFFAOYSA-N
11-azidoundecyl(triethoxy)silane
46779111
95%
Moisture sensitive
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

RUO – Research Use Only