accessibility menu, dialog, popup

UserName

H3COSiNNNOCH3OCH3

CAS RN 663171-33-3

IUPAC Name: (11-azidoundecyl)triethoxysilane
Synonyms: (11-azidoundecyl)triethoxysilane
Molecular Weight (g/mol): 359.59
Molecular Formula: C17H37N3O3Si
InChi Key: WCDFLYKXACDAES-UHFFFAOYSA-N
SMILES: CCO[Si](CCCCCCCCCCCN=[N+]=[N-])(OCC)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results