missing translation for 'onlineSavingsMsg'
Learn More

11-Azidoundecyltriethoxysilane, 95%

CAS: 663171-33-3 | C17H37N3O3Si | 359.586 g/mol

Supplier:  Thermo Scientific Chemicals H3352403

 View more versions of this product

Catalog No. AAH3352403


Only null left
Explore available promotions

Description

Description

This coupling agent specifically reacts with alkyne functions to form the corresponding 1,2,3-triazole via a copper-free 1,3-dipolar cycloaddition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This coupling agent specifically reacts with alkyne functions to form the corresponding 1,2,3-triazole via a copper-free 1,3-dipolar cycloaddition.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

110°C to 115°C (0.04mmHg)
663171-33-3
C17H37N3O3Si
MFCD11982879
WCDFLYKXACDAES-UHFFFAOYSA-N
11-azidoundecyl(triethoxy)silane
46779111
95%
Moisture sensitive
11-Azidoundecyltriethoxysilane
1 g
N3(CH2)11Si(OCH2CH3)3
azidoundecyltriethoxysilane, 11-azidoundecyltriethoxysilane, 11-azidoundecyl triethoxysilane
CCO[Si](CCCCCCCCCCCN=[N+]=[N-])(OCC)OCC
359.586
359.58
1.4390-1.4430

Chemical Identifiers

663171-33-3
359.586
WCDFLYKXACDAES-UHFFFAOYSA-N
46779111
CCO[Si](CCCCCCCCCCCN=[N+]=[N-])(OCC)OCC
C17H37N3O3Si
MFCD11982879
azidoundecyltriethoxysilane, 11-azidoundecyltriethoxysilane, 11-azidoundecyl triethoxysilane
11-azidoundecyl(triethoxy)silane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only