missing translation for 'onlineSavingsMsg'
Learn More

10-Undecynoic Acid 98.0+%, TCI America™

Supplier:  TCI America U00545G

 View more versions of this product

Catalog No. U00545G

Only null left

Description

Description

Specifications

Specifications

10-Undecynoic Acid
43°C
180°C
MFCD00014389
10-undecynoic acid, hendecynoic acid, unii-2f79g7h1wy, undec-10-ynoicacid, acmc-1cp9s, undec-1-yn-11-oic acid, 4-02-00-01738 beilstein handbook reference
C#CCCCCCCCCC(=O)O
182.263
182.26
Crystalline Powder
2777-65-3
White-Yellow
C11H18O2
5 g
OAOUTNMJEFWJPO-UHFFFAOYSA-N
undec-10-ynoic acid
31039
≥98.0% (GC)

Chemical Identifiers

2777-65-3
182.263
OAOUTNMJEFWJPO-UHFFFAOYSA-N
31039
C#CCCCCCCCCC(=O)O
C11H18O2
MFCD00014389
10-undecynoic acid, hendecynoic acid, unii-2f79g7h1wy, undec-10-ynoicacid, acmc-1cp9s, undec-1-yn-11-oic acid, 4-02-00-01738 beilstein handbook reference
undec-10-ynoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : YQ3683000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions