missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10-Undecynoic Acid 98.0+%, TCI America™
$53.16 - $178.48
Chemical Identifiers
| CAS | 2777-65-3 |
|---|---|
| Molecular Formula | C11H18O2 |
| Molecular Weight (g/mol) | 182.263 |
| MDL Number | MFCD00014389 |
| InChI Key | OAOUTNMJEFWJPO-UHFFFAOYSA-N |
| Synonym | 10-undecynoic acid, hendecynoic acid, unii-2f79g7h1wy, undec-10-ynoicacid, acmc-1cp9s, undec-1-yn-11-oic acid, 4-02-00-01738 beilstein handbook reference |
| PubChem CID | 31039 |
| IUPAC Name | undec-10-ynoic acid |
| SMILES | C#CCCCCCCCCC(=O)O |
Chemical Identifiers
| 2777-65-3 | |
| 182.263 | |
| OAOUTNMJEFWJPO-UHFFFAOYSA-N | |
| 31039 | |
| C#CCCCCCCCCC(=O)O |
| C11H18O2 | |
| MFCD00014389 | |
| 10-undecynoic acid, hendecynoic acid, unii-2f79g7h1wy, undec-10-ynoicacid, acmc-1cp9s, undec-1-yn-11-oic acid, 4-02-00-01738 beilstein handbook reference | |
| undec-10-ynoic acid |
Specifications
| 2777-65-3 | |
| White-Yellow | |
| C11H18O2 | |
| 1 g | |
| OAOUTNMJEFWJPO-UHFFFAOYSA-N | |
| undec-10-ynoic acid | |
| 31039 | |
| ≥98.0% (GC) | |
| 10-Undecynoic Acid |
| 43°C | |
| 180°C | |
| MFCD00014389 | |
| 10-undecynoic acid, hendecynoic acid, unii-2f79g7h1wy, undec-10-ynoicacid, acmc-1cp9s, undec-1-yn-11-oic acid, 4-02-00-01738 beilstein handbook reference | |
| C#CCCCCCCCCC(=O)O | |
| 182.263 | |
| 182.26 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : YQ3683000
TSCA : No