accessibility menu, dialog, popup

UserName

CAS RN 2777-65-3

IUPAC Name: undec-10-ynoate
Synonyms: undec-10-ynoate
Molecular Weight (g/mol): 181.26
Molecular Formula: C11H17O2
InChi Key: OAOUTNMJEFWJPO-UHFFFAOYSA-M
SMILES: [O-]C(=O)CCCCCCCCC#C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)