Learn More
10-(Pentafluorophenoxycarbonyl)decyltriethoxysilane, 95%
CAS: 1197981-08-0 | C23H35F5O5Si | 514.605 g/mol
$211.84 - $575.49
Chemical Identifiers
| CAS | 1197981-08-0 |
|---|---|
| Molecular Formula | C23H35F5O5Si |
| Molecular Weight (g/mol) | 514.605 |
| MDL Number | MFCD13181916 |
| InChI Key | WQFFEEIHQJVCHR-UHFFFAOYSA-N |
| Synonym | 2,3,4,5,6-pentafluorophenyl 11-triethoxysilyl undecanoate, 10-pentafluorophenoxycarbonyl decyltriethoxysilane, 10-pentafluorophenoxycarbonyl decyl triethoxysilane |
| PubChem CID | 99738205 |
| IUPAC Name | (2,3,4,5,6-pentafluorophenyl) 11-triethoxysilylundecanoate |
| SMILES | CCO[Si](CCCCCCCCCCC(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)(OCC)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH34037MD
|
Thermo Scientific Chemicals
H34037MD |
250 mg |
Each for $211.84
|
|
|||||
|
AAH3403703
|
Thermo Scientific Chemicals
H3403703 |
1 g |
Each for $575.49
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1197981-08-0 | |
| 514.605 | |
| WQFFEEIHQJVCHR-UHFFFAOYSA-N | |
| 99738205 | |
| CCO[Si](CCCCCCCCCCC(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)(OCC)OCC |
| C23H35F5O5Si | |
| MFCD13181916 | |
| 2,3,4,5,6-pentafluorophenyl 11-triethoxysilyl undecanoate, 10-pentafluorophenoxycarbonyl decyltriethoxysilane, 10-pentafluorophenoxycarbonyl decyl triethoxysilane | |
| (2,3,4,5,6-pentafluorophenyl) 11-triethoxysilylundecanoate |
Specifications
| 155°C to 160°C (0.04mmHg) | |
| 250 mg | |
| MFCD13181916 | |
| WQFFEEIHQJVCHR-UHFFFAOYSA-N | |
| (2,3,4,5,6-pentafluorophenyl) 11-triethoxysilylundecanoate | |
| 99738205 | |
| 95% | |
| Moisture sensitive |
| 1197981-08-0 | |
| C23H35F5O5Si | |
| 2,3,4,5,6-pentafluorophenyl 11-triethoxysilyl undecanoate, 10-pentafluorophenoxycarbonyl decyltriethoxysilane, 10-pentafluorophenoxycarbonyl decyl triethoxysilane | |
| CCO[Si](CCCCCCCCCCC(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)(OCC)OCC | |
| 514.605 | |
| 514.61 | |
| 1.4320-1.4360 | |
| 10-(Pentafluorophenoxycarbonyl)decyltriethoxysilane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only