missing translation for 'onlineSavingsMsg'
Learn More

10-(Pentafluorophenoxycarbonyl)decyltriethoxysilane, 95%

CAS: 1197981-08-0 | C23H35F5O5Si | 514.605 g/mol

Supplier:  Thermo Scientific Chemicals H3403703

 View more versions of this product

Catalog No. AAH3403703


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

155°C to 160°C (0.04mmHg)
1197981-08-0
C23H35F5O5Si
2,3,4,5,6-pentafluorophenyl 11-triethoxysilyl undecanoate, 10-pentafluorophenoxycarbonyl decyltriethoxysilane, 10-pentafluorophenoxycarbonyl decyl triethoxysilane
CCO[Si](CCCCCCCCCCC(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)(OCC)OCC
514.605
514.61
1.4320-1.4360
10-(Pentafluorophenoxycarbonyl)decyltriethoxysilane
1 g
MFCD13181916
WQFFEEIHQJVCHR-UHFFFAOYSA-N
(2,3,4,5,6-pentafluorophenyl) 11-triethoxysilylundecanoate
99738205
95%
Moisture sensitive

Chemical Identifiers

1197981-08-0
514.605
WQFFEEIHQJVCHR-UHFFFAOYSA-N
99738205
CCO[Si](CCCCCCCCCCC(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)(OCC)OCC
C23H35F5O5Si
MFCD13181916
2,3,4,5,6-pentafluorophenyl 11-triethoxysilyl undecanoate, 10-pentafluorophenoxycarbonyl decyltriethoxysilane, 10-pentafluorophenoxycarbonyl decyl triethoxysilane
(2,3,4,5,6-pentafluorophenyl) 11-triethoxysilylundecanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only