accessibility menu, dialog, popup

UserName

H3COSiOOFFFFFOH3COCH3

CAS RN 1197981-08-0

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 11-(triethoxysilyl)undecanoate
Synonyms: 2,3,4,5,6-pentafluorophenyl 11-(triethoxysilyl)undecanoate
Molecular Weight (g/mol): 514.61
Molecular Formula: C23H35F5O5Si
InChi Key: WQFFEEIHQJVCHR-UHFFFAOYSA-N
SMILES: CCO[Si](CCCCCCCCCCC(=O)OC1=C(F)C(F)=C(F)C(F)=C1F)(OCC)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results