missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 10-Hydroxycamptothecin

Catalog No. AC467490050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467490050 5 g
AC467490010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467490050 Supplier Thermo Scientific Chemicals Supplier No. 467490050

missing translation for 'validMainframeMsg'

CAS: 19685-09-7 | C20H16N2O5, C20H16N2O5 | 364.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19685-09-7
Molecular Formula C20H16N2O5

Specifications

Quantity 5 g
Melting Point 225.0°C to 267.0°C
Formula Weight 364.36
Chemical Name or Material 10-Hydroxycamptothecin

RUO – Research Use Only