missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+/-)-10-Camphorsulfonic Acid 98.0+%, TCI America™
$51.26 - $391.11
Chemical Identifiers
| CAS | 8-2-5872 |
|---|---|
| Molecular Formula | C10H16O4S |
| Molecular Weight (g/mol) | 232.29 |
| MDL Number | MFCD00074827 |
| InChI Key | MIOPJNTWMNEORI-UHFFFAOYNA-N |
| Synonym | reychler's acid, camphorsulfonic acid, d-camphorsulfonic acid, camphersulfosaeure, --10-camphorsulfonic acid, 2-oxobornane-10-sulphonic acid, d-10-camphorsulfonic acid, 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, l-camphor-10-sulfonic acid, +-10-camphorsulfonic acid |
| PubChem CID | 18462 |
| ChEBI | CHEBI:55379 |
| IUPAC Name | (7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonic acid |
| SMILES | CC1(C2CCC1(C(=O)C2)CS(=O)(=O)O)C |
Chemical Identifiers
| 8-2-5872 | |
| 232.29 | |
| MIOPJNTWMNEORI-UHFFFAOYNA-N | |
| 18462 | |
| (7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonic acid |
| C10H16O4S | |
| MFCD00074827 | |
| reychler's acid, camphorsulfonic acid, d-camphorsulfonic acid, camphersulfosaeure, --10-camphorsulfonic acid, 2-oxobornane-10-sulphonic acid, d-10-camphorsulfonic acid, 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, l-camphor-10-sulfonic acid, +-10-camphorsulfonic acid | |
| CHEBI:55379 | |
| CC1(C2CCC1(C(=O)C2)CS(=O)(=O)O)C |
Specifications
| 8-2-5872 | |
| White-Yellow | |
| MFCD00074827 | |
| reychler's acid, camphorsulfonic acid, d-camphorsulfonic acid, camphersulfosaeure, --10-camphorsulfonic acid, 2-oxobornane-10-sulphonic acid, d-10-camphorsulfonic acid, 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, l-camphor-10-sulfonic acid, +-10-camphorsulfonic acid | |
| CC1(C2CCC1(C(=O)C2)CS(=O)(=O)O)C | |
| 232.29 | |
| CHEBI:55379 | |
| ≥98.0% (T) | |
| (+/-)-10-Camphorsulfonic Acid |
| 202°C | |
| C10H16O4S | |
| 3261 | |
| MIOPJNTWMNEORI-UHFFFAOYNA-N | |
| (7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonic acid | |
| 18462 | |
| 232.29 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0309
TSCA : Yes